Order Entry
Germany
ContactUsLinkComponent
VWR®, Thiocyanate 1000 mg/L in Water standard for ion chromatography
VWR®, Thiocyanate 1000 mg/L in Water standard for ion chromatography
Catalog # 84999.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Thiocyanate 1000 mg/L in Water standard for ion chromatography
Catalog # 84999.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Thiocyanate 1,000 mg/l in water standard for ion chromatography
  • Contents:
    NH₄SCN in H₂O

Specifications

About this item

Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.