Catheter syringes
Supplier: SEMADENI
Certificates
About this item
These catheter syringes have a PP barrel with silicone rubber piston.
- Can be uesd as suction and irrigation syringes
- Also used in technical purposes for pastes, glues and more
These syringes are gas sterilisable.
Packaging: Individually packed.