Order Entry
Germany
ContactUsLinkComponent
Desiccator cabinet, SICCO wall
Desiccator cabinet, SICCO wall
Catalog # BOHLV1994-08
Supplier:  Bohlender
undefined
Desiccator cabinet, SICCO wall
Catalog # BOHLV1994-08
Supplier:  Bohlender

Specifications

  • W×D×H ext.:
    1080×580×1870 mm
  • Pk:
    1
  • Shelves:
    12
  • W×D×H int.:
    495×540×500 mm

Specifications

About this item

Transparent PPMA cabinet with reinforced aluminium frame, unit has a door with a foam rubber gasket and magnetic strip. Designed for storing or drying humidity sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA-samples.

  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Temperature resistance from −20 to +70 °C
  • Easy to move about as unit has four castors, two of them with brakes
Delivery: Unit has six compartments, each with its own door, removable stainless steel telescopic shelf, desiccant tray and hygrometer. Each chamber can accommodate maximum of 17 shelves.