Cleanroom bond paper, EcoBond®
Supplier: BERKSHIRE
Certificates
About this item
EcoBond® is a recyclable version of BCR® cleanroom bond paper. EcoBond® is a made from cellulose and is recommended for use in the ISO Class 3 to 7 cleanrooms.
- Low in particles and fibres
- Double-packed for cleanliness
- Recyclable
- Smooth surfaces reduce particle contamination
Related Information
Certificates
Search by Lot NumberEcoBond® Recyclable Cleanroom Bond Paper