Antiseptic spray, Cutasept® med F
148-4191
: Bode
- Description:Antiseptic spray with ethanol
- Pk:250 mL
Colourless antiseptic spray with ethanol, for use prior to injections, punctures and surgical procedures.
- Ideal for use prior to blood glucose monitorings and insulin administration
- Excellent skin compactibility, hypoallergenic
- Effects: Bactericidal, yeasticidal, virucidal against eneveloped viruses (HBV, HIV, HCV)
- Convenient invertible spray pump without propellants
Exposure time: 15 sec to 2 min