Order Entry
Germany
ContactUsLinkComponent
Detergent, concentrated, Shureclean Plus VK9
Detergent, concentrated, Shureclean Plus VK9
Catalog # DIVY7509854
Supplier:  Diversey
Detergent, concentrated, Shureclean Plus VK9
Catalog # DIVY7509854
Supplier:  Diversey

Specifications

  • Description:
    Detergent, Shureclean Plus VK9, concentrated, neutral and high foaming
  • Seq:
    01
  • Packed:
    2×5 l
  • Pk:
    2 Can

Specifications

About this item

Shureclean Plus VK9 is a concentrated, neutral, high foaming liquid detergent designed for general purpose applications in the food, beverage and dairy industries. This detergent is recommended for manual and soak cleaning of all food preparation equipment, cooking vessels and utensils. It can also be used for cleaning floors, walls and work surfaces.

  • Clear, pale brown liquid
  • Relative density at 20 °C - 1,03
  • Suitable for soft or hard water and pH neutral
  • Non perfumed product