ContactUsLinkComponent
Surgical masks, Foliodress® mask comfort perfect
Surgical masks, Foliodress® mask comfort perfect
Catalog # HART992143/1
Supplier:  HARTMANN PAUL
undefined
Surgical masks, Foliodress® mask comfort perfect
Catalog # HART992143/1
Supplier:  HARTMANN PAUL
Specifications
  • Type:
    With ties
  • Colour:
    Green
  • Pk:
    50
Specifications
About this item
Comfortable surgical face masks made from hypoallergenic non-woven materials.
  • Integrated filter for a high bacterial filtration efficiency and low breathing resistance
  • Long integrated nosepiece
  • With ultrasonically sealed ties
  • Free from natural rubber latex, fragrance free and durable
Suitable for all surgical areas sucha as isolation wards, intensive care departments, delivery rooms, premature and newborn infant wards.
Certifications: EN 14683 Type II