Surgical masks, Foliodress® Mask Comfort Senso
HART9921381
- Type:With ties
- Colour:Green
- Pk:50
Comfortable surgical face masks made from special combination of extra soft, lightweight and hypoallergenic non-woven materials.
- For wearers with sensitive skin
- Integrated filter for a high bacterial filtration efficiency and low breathing resistance
- Long integrated nosepiece
- Ultrasonically sealed ties
- Free from natural rubber latex, fragrance free and durable
Suitable for use in all surgical areas, like isolation wards, intensive care departments, delivery rooms, premature and newborn infant wards.
: EN 14683 Type II