Hand disinfectants, Sterillium® Virugard
Supplier: Bode
Certificates
About this item
Sterillium® Virugard is a ethanol based, highly effective virucidal hand disinfectant. Particularly suitable for all areas with an increased risk of infection.
- Particularly active against enveloped and nonenveloped viruses
- Bactericidal, yeasiticdal, fungicidal, tuberculocidal, mycobactericidal and virucidal
- Recommended by the RKI for epidemic outbreaks including the noroviruses
- Colourant- and fragrance-free
Caution: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.
* These articles are only available in Germany.