Hand disinfectants, Sensiva®
Catalog # SCUE115203
Supplier: SCHÜLKE & MAYR GMBH
Specifications
- Description:Hand disinfectant, Sensiva®, 1000 ml dispenser bottle
- Pk:10
Specifications
About this item
Sensiva® hand-disinfection is an alcohol-based, rub-in product for hygienic and surgical hand disinfection.
- Moisture-retaining factors and moisturesing agents prevents skin degraesing and dehydration
- Active against bacteria, skin fungi and lipophilic viruses (HBV, HIV, vaccinia viruses)
- Coulorant- and fragrance-free