Hand disinfectant gels, ANIOSGEL 85
Supplier: Laboratoires Anios
Certificates
About this item
Thixotrophic hydroalcoholic gel for hygienic treatment and surgical dinfection by hand rubbing.
- Active against bacteria, yeasts, moulds and viruses
- Hygenic treatment: 3 ml for 30 s
- Can be used without water
- Colourant- and fragrant-free
Caution: Use biocides safely. Always read the label and product information before use.