ContactUsLinkComponent
Disinfectant products, Korsolex® extra
Disinfectant products, Korsolex® extra
Catalog # BODE980254
Supplier:  BODE CHEMIE
undefined
Disinfectant products, Korsolex® extra
Catalog # BODE980254
Supplier:  BODE CHEMIE
Specifications
  • Description:
    Korsolex® extra
  • Seq:
    01
  • Packed:
    2 litre bottle
  • Pk:
    4
Specifications
About this item
Korsolex® extra is a broad spectrum disinfectant based on glutaraldehyde and quaternary ammonium compounds and is designed for heat-sensitive and heat-resistant instruments. Korsolex® extra is suitable for all common manual and semi-automatic circulation procedures as well as for the fully automatic (cold disinfection) procedure.
  • Virucidal
  • User friendly with excellent material compatibility
  • Suitable in combination with chemothermal treatment
  • Typical dosage: 5% in 5 minutes
Certifications: Bactericidal: EN 14561, EN 13727; Fungicidal EN 1275, EN 13624, EN 14562; Mycobatericidal: EN 14348; Virucidal: EN 14476.