Outdoor sealing mat, for drains
: asecos
Sealing mats made from a dense and flexible material prevent fast and efficient penetration of water-polluting liquids into the ground or drains.
- Rapid protection against spilled liquids
- Resistant to hydrocarbon and most chemicals
- Reusable, can be washed of with soap and water
Colour: White/yellow