Wipes, pre-saturated
Supplier: ESSENTRA POROUS TECHNOLOGIES
Certificates
About this item
These heavyweight polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
- Pre-wetted with high concentration of alcohol
- Individually packed: Easy tear, single use foil package
- Dissolves most residues safely and effectively
- Excellent as a degreaser
- Made in the USA
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904