Water-proof shoe protection spray
Catalog # UVEX9698.100
Supplier: UVEX
Specifications
- Description:Water-proof shoe protection spray, 100 ml
- Seq:01
- Pk:1
Specifications
About this item
This waterproof and stain-resistant spray for shoes is manufactured by using nanotechnology and chemical nanotechnology modified surfaces.
- Breathability of the shoe is fully retained
- Haptic properties are fully preserved
- Long-term effect and without propellants
Suitable for textiles and leather products.