Hybridisation bottles
Hybridisation bottles, GL 45 screw cap with silicone sealing, 38×150 mm, Hybridisation ovens type Hybaid, Bellco, Labline, GFL, Heraeus
WITG2.446.115
Borosilicate glass 3.3, with GL 45 screw cap and silicone sealing.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Ø×L:38×150 mm
- For:Hybridisation ovens type Hybaid, Bellco, Labline, GFL, Heraeus
- Description:Hybridisation bottles, GL 45 screw cap with silicone sealing