Orders LinkContactUsLinkComponent
Heating tapes, HST
Supplier:   HORST
Heating tapes, HST
Supplier:  HORST
Certificates
Heating tapes, HST
7 Options Available Below
Product Details & Documents
These slim heating tapes consist of two PTFE insulated heating conductors with earthed protection braid which have a common coating made of metal cloth.
Product Family Options
Product Information
  • Availability
    Price
  • HORS025201
    Heating tape HST with 0,5 m length, 30 W output
    Heating tape HST with 0,5 m length, 30 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,00 €
    81,00 €
    1 * 1 ST
       81,00 €
  • HORS025202
    Heating tape HST with 1 m length, 50 W output
    Heating tape HST with 1 m length, 50 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,00 €
    96,00 €
    1 * 1 ST
       96,00 €
  • HORS025203
    Heating tape HST with 2 m length, 100 W output
    Heating tape HST with 2 m length, 100 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • HORS025204
    Heating tape HST with 3 m length, 150 W output
    Heating tape HST with 3 m length, 150 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €
  • HORS025205
    Heating tape HST with 4 m length, 200 W output
    Heating tape HST with 4 m length, 200 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €
  • HORS025206
    Heating tape HST with 5 m length, 250 W output
    Heating tape HST with 5 m length, 250 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 214,00 €
    214,00 €
    1 * 1 ST
       214,00 €
  • HORS025207
    Heating tape HST with 10 m length, 480 W output
    Heating tape HST with 10 m length, 480 W output
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 368,78 €
    368,78 €
    1 * 1 ST
       368,78 €