Orders LinkContactUsLinkComponent
Safety Container
Safety Container
Transparent, Test tube racks or small vessels with bacteriological or hazardous substances, 330×175×180 mm, 300×125×160 mm
Catalog # SEMA3651
Supplier:   SEMADENI
Safety Container
Catalog # SEMA3651
Supplier:  SEMADENI
Safety Container
Safety Container
Transparent, Test tube racks or small vessels with bacteriological or hazardous substances, 330×175×180 mm, 300×125×160 mm
Price
264,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Safety container made of PC, with hermetically sealed lid and stainless steel mounting bracket.
  • Robust design
  • Avoid direct contact with solvents, strong acids, and alkaline solutions
Ideal for the safe storage and transport of test tube racks and small vessels with bacteriological and hazardous substance.
Specifications
  • For:
    Test tube racks or small vessels with bacteriological or hazardous substances
  • W×D×H ext.:
    330×175×180 mm
  • Colour:
    Transparent
  • W×D×H int.:
    300×125×160 mm