ContactUsLinkComponent
Container with Wooden Pallet
Container with Wooden Pallet
Catalog # SEMA2082
Supplier:  SEMADENI
undefined
Container with Wooden Pallet
Catalog # SEMA2082
Supplier:  SEMADENI
Specifications
  • Capacity:
    1000 l
  • W×D×H:
    1200×1000×1170 mm
  • Pk:
    1
Specifications
About this item
HDPE, natural
  • UV stabilised for storage outside
  • Robust tubular steel frame provides additional protection during transport
  • Sealable in- and outlet
Space-saving and transportable container for fluids and chemicals.