Order Entry
Germany
ContactUsLinkComponent
Double bossheads
Double bossheads
Catalog # BOCH5342
Supplier:  BOCHEM
undefined
Double bossheads
Catalog # BOCH5342
Supplier:  BOCHEM

Specifications

  • Material:
    Aluminium
  • Pk:
    1
  • For rod Ø:
    16,5 mm

Specifications

About this item

Double bossheads made of aluminium.

  • For rods up to 16,5 mm
  • Angle: 90°
  • With socket screw (M6)