Orders LinkContactUsLinkComponent
Clamps
Clamps
Supplier:   BOCHEM
Clamps
Supplier:  BOCHEM
Certificates
Clamps
4 Options Available Below
Product Details & Documents
Retort clamps made of stainless steel.
  • Jaws are covered with a layer of cork to protect clamped items
  • Angled jaws
Certifications: DIN 12894
Product Family Options
Product Information
  • Material
    Spanning width
    Length
    Availability
    Price
  • Stainless steel 18/10
    25 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,40 €
    22,40 €
    1 * 1 ST
       22,40 €
  • Stainless steel 18/10
    40 mm
    150
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,40 €
    24,40 €
    1 * 1 ST
       24,40 €
  • Stainless steel 18/10
    60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,40 €
    27,40 €
    1 * 1 ST
       27,40 €
  • Stainless steel 18/10
    80 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,70 €
    35,70 €
    1 * 1 ST
       35,70 €