Cut resistant gloves, Check & Go Green PU 5
Supplier: HONEYWELL SAFETY
Certificates
About this item
These knitted gloves in polyamide/HPPE fibres/elastane and glass fibres (outside) feature a green PU coating on the palm. Ideal for handling sharp parts in an oily or greasy environment.
- Tight knit (gauge 13) gives the glove perfect support, limits penetration of dirt and offers a good sense of touch
- Ventilated back optimises breathability
- PU coating of the palm and fingertips provides outstanding abrasion resistance and oil protection
- Composite fibre fully protected by polyamide increases the cut resistance
- Colour-coding on top of the hand and coating colour helps safety manager and workers to visually recognise the level of cut protection
Typical application areas include: Automotive and part manufacturing, aviation building and construction, catering, energy or electricity, glass industries, metal and paper industries, logistics and maintenance.
Gauge: 13
Colour: White/green
EN 388 (4543); Cat. II
Certifications: EN 388:2016 (3X43C); Cat. II
Specifications
- Catalog No:
- 112-4656
- 112-4658
- 112-4655
- 112-4657
- 112-4654
- Model:
- PU 5
- PU 5
- PU 5
- PU 5
- PU 5
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Finish:
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)