VWR® SeraFree, Cryopreservation Media
Supplier: VWR Chemicals
Certificates
About this item
SeraFree cryopreservation media is ready to use freezing media for cryopreservation of adherent or suspension cultured cells. The animal-free media composition eliminates batch-to-batch variability and is optimised for cell viability and cell growth after thawing. Suitable for use in environments that prohibit the use of animal-derived products.
Endotoxin tested
- Ready to use, animal-free, RPMI- or DMEM-based media
- Optimises cell growth and cell viability after thawing
- Reduces potential for transmission of infectious diseases