VWR® B'Paper, Cleanroom paper
Supplier: VWR Collection
Certificates
About this item
Formulated with coated paper, without magnesium silicate, b-paper 80 or 92 g is cut, cleaned and packed in a Class 100 (ISO 5) cleanroom.
- Different colours available
- Can be used with inkjet or laser printers, with copy machines, and is also suitable for note taking
- Low ionic and particle contamination
- Ideal for all cleanroom applications