Cleanroom wipes, CleanWIPE®
Supplier: FOAMTEC
Certificates
About this item
These soft, nonabrasive wipers trap particulate matter in a skeletal structure designed to reduce particulate contamination.
- Allow rapid absorption of spillage with superior wet tensile strength
- Superior solvent applicators with sponge-like qualities
- Thickness of 2,76 mm
These wipers used for removal of alcohol or amyl acetate, for spills, and for wiping desk surfaces and workstations. Also suitable for thick film screen cleaning. Suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanrooms.
Certified cytotoxic free. Wipers are laundered with a proprietary process yielding a Class 100 wiper that meets rigorous cytotoxicity testing for the medical device industry and these are double bagged.
Certifications: Particulate requirement counts are certified in accordance with ASTM F-51-65T for ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.