ContactUsLinkComponent
Sample cups, PelletCups® and PlastiCup™
Sample cups, PelletCups® and PlastiCup™
Catalog # CHPXCHX-554
Sample cups, PelletCups® and PlastiCup™
Catalog # CHPXCHX-554
Specifications
  • Ext.Ø:
    39,7 mm
  • Packed:
    Bag of 1000
  • Type:
    PlastiCup™ compressible tapered plastic briquetting cup
  • Height:
    6,4 mm
  • Material:
    Plastic
  • Seq:
    07
  • Pk:
    500