Orders LinkContactUsLinkComponent
Safety spectacles, SecureFit™ 200 | Avantor
Safety spectacles, SecureFit™ 200
Supplier:   3M
Safety spectacles, SecureFit™ 200
Supplier:  3M
Zertifikate
Safety spectacles, SecureFit™ 200
Safety spectacles, SecureFit™ 200
2 mögliche Produktvariationen vorhanden
Product Details & Documents
Stylish, lightweight designed safety glasses features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • 3M Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes
  • Self-adjusting feature eliminates need to stock multiple frame sizes at worksites
  • Secure, snug fit keeps eyewear in place without slippage even when the person is in motion
Weight: 18 g
Zertifizierung: EN 166.
Product Family Options
Produktinformationen
  • Type
    Colour
    Lenses
    Availability
    Price
  • Anti-UV, anti-scratch, anti-fog
    Grey
    Grey PC
    Anmelden für die Verfügbarkeit
    Anmelden für die Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
  • Anti-UV, anti-scratch, anti-fog
    Transparent
    Clear PC
    Anmelden für die Verfügbarkeit
    Anmelden für die Verfügbarkeit
    1 * 1 ST
    1 * 1 ST