Order Entry
Germany
ContactUsLinkComponent
VWR®, Wipes, Pre-Saturated
VWR®, Wipes, Pre-Saturated
Catalog # 115-0128
Supplier:  VWR Collection
undefined
VWR®, Wipes, Pre-Saturated
Catalog # 115-0128
Supplier:  VWR Collection

Specifications

  • Packed:
    100 wipes/container
  • L×W:
    152×228 mm
  • Seq:
    03
  • Pk:
    100

Specifications

About this item

These non woven wipes are constructed from 100% polypropylene and are moistened with a blend of pure, reagent grade 70% isopropyl alcohol and 30% reagent grade deionised water. They are ideal for wiping optical parts, delicate glassware, electrodes, cuvettes, microscopes or lenses.

  • Suitable for all applications in environmentally-sensitive areas
  • Very strong, soft and lint-free
  • Will not scratch surfaces
  • Individually dispensed wipes are handy and ready for instant use
  • Reclosable bench top container ensures a clean wipe every time, snap-on sealed top eliminates contamination from dust or dirt
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904