ContactUsLinkComponent
Adhesive remover, spray, Scotch-Weld™
Adhesive remover, spray, Scotch-Weld™
Catalog # MMMAIR500
Supplier:  3M
undefined
Adhesive remover, spray, Scotch-Weld™
Catalog # MMMAIR500
Supplier:  3M
Specifications
  • Description:
    Scotch-Weld™ cleaner spray
  • Pk:
    500 mL
Specifications
About this item
Powerful citrus-based adhesive remover spray cleaner effectively removes adhesive residue, grease and tar.
  • Clean and degrease metal parts, motors, tools, printing presses and concrete
  • Clean annoying residue from sticky labels
  • Aerosol spray can allows quick and easy application
  • Pleasant citrus smell