Order Entry
Germany
ContactUsLinkComponent
Syringes, 3-piece, BD Emerald™
Syringes, 3-piece, BD Emerald™
Catalog # BDAM307727UK
Supplier:  BD Medical
undefined
Syringes, 3-piece, BD Emerald™
Catalog # BDAM307727UK
Supplier:  BD Medical

Specifications

  • Capacity:
    2 ml
  • Description:
    Syringe without needle, Luer slip, centric tip
  • Seq:
    01
  • Graduation:
    0,1 ml
  • Pk:
    3.000

Specifications

About this item

Sterile, disposable, three-piece, PP barrel and piston, latex-free stopper. Luer slip syringes available with or without needle.

  • Clear barrel and green stopper for perfect visibility of syringe contents
  • Special silicone lubrication ensures that plunger moves smoothly and evenly
  • Leakproof even under high pressure
  • Sterilised by ethylene oxide
Packaging: Individually packed in blister film and medical grade paper.