ContactUsLinkComponent
Standard tube holders, VACUETTE®
Catalog # GREI450209_100
Supplier:  Greiner Bio-One
Standard tube holders, VACUETTE®
Catalog # GREI450209_100
Supplier:  Greiner Bio-One
Specifications
  • Description:
    Plastic, short version, non-sterile
  • Seq:
    03
  • Pk:
    100
Specifications
About this item
These VACUETTE® standard tube holders are non-sterile and made of plastic.
  • Special, user-friendly design
  • Good grip when drawing blood due to structured surface
  • Safer collection process
They are compatible with all VACUETTE® needles, VACUETTE® Luer™ adapters and blood collection sets. The use of these products with needles from other manufacturers is the responsibility of the user.