Capillary columns, traditional phases, SLB-5ms, Supelco®
: Merck
This column provides boiling point elution order with a slight increase in selectivity, especially for aromatic compounds.
- Bonded and highly crosslinked
- High thermal stability - maximum temperatures up to −60 to 330° C
- Low bleed even at elevated temperatures