Order Entry
Germany
ContactUsLinkComponent
Trays, styrene acrylonitrile
Trays, styrene acrylonitrile
Catalog # 216-7310
Supplier:  SEMADENI
undefined
Trays, styrene acrylonitrile
Catalog # 216-7310
Supplier:  SEMADENI

Specifications

  • Colour:
    White
  • W×D×H:
    420×280×20 mm
  • Pk:
    1

Specifications

About this item

Trays made of styrene acrylonitrile, white.

  • Temperature resistance up to 85 °C
  • Easy to clean

Ideal for use as a serving tray.