Wafer tweezers, Erem®
Supplier: Basan
Certificates
About this item
Stainless steel. These tweezers are suitable for 4 to 6" wafers. They feature a flat lower paddle and either 6 upper fingers or a free-floating Teflon® upper paddle.
- Serrated finger grips for secure handling
- Anti-magnetic, non rusting
- Acid-proof, heat resistant surface
- Model with 6 upper fingers for protecting wafers against damage
- Model with free-floating Teflon upper paddle for secure, damage-free gripping