ContactUsLinkComponent
Actril Fog System (AFS)
Actril Fog System (AFS)
  MINN3025825
Actril Fog System (AFS)
  MINN3025825
  • Description:
    Actril® fog system
  • Pk:
    1
Fogging technology using Actril® Cold Sterilant ready-to-use product naturally disinfects surfaces through vapour condensation for multiple applications: hospitals, animal facilities, schools and locker rooms....
  • High level bio-decontamination, fast microbiological efficacy
  • Destroys bacteria, mycobacteria, yeast, mold, virus and bacteria spores
  • Environmentally friendly
  • No manual spraying or wiping, no human contact with chemical aerosols, no residue left on surfaces
  • Multiple Applications
  • Low investment, no maintenance, minimal chemical usage
The Actril® Fog dispenser is a specially designed device that fits on 1 gallon and 5 L Actril® containers to produce an Actril® vapor which coats and disinfects any hard surface. The device fits on the top of a standard Actril® bottle and uses an 8 mm or ⁵/₁₆" diameter compressed air line supplied with compressed air (70 L/min @ 3 bar or 2,5 SCFM @ 45 PSI) to fog the Actril® Cold Sterilant from the Actril Fog System.
The Actril® re-condensates on hard surfaces and destroys the exisiting micro-organisms. The fog system includes the fogging dispenser and a bottle cradle to prevent the bottle from tipping over during fogging. The sytem is capable of disinfecting room volumes up to 100 m³.