PCR plates, 96-well
Supplier: Brand
Certificates
About this item
PP.
- Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
- Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
- Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
- Free from DNase, RNase and DNA
- Autoclavable at 121 °C (2 bar), acc. DIN EN 285
96-well non skirted PCR plates are suitable for most commercially available thermal cyclers. Plates have blue alphanumeric code and cut corner marking.
96-well semi-skirted PCR plates can easily be labelled or tagged with a barcode. Plates have blue alphanumeric code and cut corner marking.
96-well skirted PCR plates are especially rigid, and are optimally suited for use with automatic pipetting systems and for being transported. Plates have black alphanumeric code, with no additional markings on the cut corner.
Related Information
Certificates
Search by Lot NumberBrand PCR Products
Product Literature
Characterisation of Antibodies with PCR Plates PCR Plates PCR System
Application Notes
Improved sealing surface of PCR plates from BRAND® to support reliable evaporation protection Protection of samples and reliable results with BRAND matching sealing options
TDS PCR Products