Orders LinkContactUsLinkComponent
Kidney dishes
260×100×55 mm
  SEMA1254
 :   SEMADENI
Kidney dishes
  SEMA1254
 :  SEMADENI
Kidney dishes
Kidney dishes
260×100×55 mm
26,90 €
1 * 1 ST
Kidney dishes made of white PP.
  • Temperature resistant up to 130 °C
  • Excellent chemical resistance
  • W×D×H:
    260×100×55 mm