IP ActivFlo Biopsy III cassettes, taped
Supplier: LEICA HISTOLOGY
Certificates
About this item
IP ActivFlo Biopsy III cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,26 mm pore size, the cassettes can secure the smallest specimens, eliminating the need for biopsy bags and sponges. The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lid have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
- Available in taped packaging for easy printer loading
- Eleven colours available