Tissue paper, Bio-Wraps
720-2430
- Colour:Blue
- Description:Bio- Wraps, biopsy paper
- Dimensions:63,5×101,6 mm
- Pk:500
Bio-Wraps are sections of tissue paper cut to a convenient size.
- They will not bleed into specimens
- Bio-Wraps are available in convenient dispenser packs
They are blue coloured to help locate small biopsy specimens following paraffin infiltration.