Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microscope Eyepieces
Bodies for Stemi 2000
Bodies for Stemi 2000
Bodies for Stemi 2000
630-0367
:
ZEISS
Bodies for Stemi 2000
630-0367
:
ZEISS
Description:
Body with apochromatic Greenough with stereoscopic angle 11°, 0,65 - 5,0× and dust protection cap with M49×0,75 for photo filter
Seq:
02
Pk:
1