Orders LinkContactUsLinkComponent
Electrode cleaning solutions
Electrode cleaning solutions
Electrode cleaning solutions
Supplier:  Mettler - Toledo
Certificates
Electrode cleaning solutions
2 Options Available Below
Product Details & Documents
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • METN51340068
    Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 1×250 ml
    Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 1×250 ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 26,61 €
    26,61 €
    1 * 250 mL
       26,61 €
  • 662-0573
    Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 6×250 ml
    Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 6×250 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 119,88 €
    119,88 €
    1 * 6 ST
       119,88 €
This product has been blocked by your organization. Please contact your purchasing department for more information