Order Entry
Germany
ContactUsLinkComponent
 
Cooling box, mini electric cooler
  478-1265
 :  NEOLAB
Cooling box, mini electric cooler
  478-1265
 :  NEOLAB

 

  • Capacity:
    21 l
  • Dimensions:
    320×240×420 mm (ext)
  • Pk:
    1

 

 

Mini electric cooler, GS tested, with CFC-free full-foam isolation for the connection on a car battery (12 V).

  • Easy to handle
  • Removable lid builted-in cooling aggregate
  • Easy to clean
 : Supplied with shoulder carrying strap.