ContactUsLinkComponent
Pocket thermometer, QUICKFIT®
Pocket thermometer, QUICKFIT®
  620-2286
undefined
Pocket thermometer, QUICKFIT®
  620-2286
  • Description:
    Pocket thermometer for Quickfit® still heads, 14/23 cone size, 45 mm
  • Pk:
    1
Quickfit® ground-glass joints are precision manufactured with an accurate taper and smooth finish for leak-free performance and assured interchangeability.
  • For incorporating thermometers into Quickfit® jointed glassware assemblies
  • Pocket fits most Quickfit® still heads