Glove liners, 265R
: Showa
These seamless knitted nylon liners with thin nitrile coating protect the hand from oils, hydrocarbons, grease and abrasion.
- Light, elastic, low-lint glove, resistant to deformation
- Breathable back reduces perspiration
- Anatomical shape: Designed for easy movement and continuous wear
- Elasticated cuffs which protect the wrist
Ideal for people working in agriculture, horticulture and construction.
Length: 240 - 270 mm
EN 388 (3131); Cat. II
: EN 388 (3131X); Cat. II.
- Catalog No:
- 112-5752
- 112-5751
- 112-5753
- 112-5754
- Model:
- 265R
- 265R
- 265R
- 265R
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (3131X)
- EN 388 (3131X)
- EN 388 (3131X)
- EN 388 (3131X)