High-purity tubing, Tygon® 2475
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Tygon®, High-purity tubing providing a high level of purity not previously available in a clear, flexible tubing.
- Exceptionally low absorption and adsorption compared to silicone
- Smooth inner surface inhibits particulate entrapment
- Environmentally safer to dispose
- Highly resistant to aggressive cleaners and sanitisers
- Plasticiser free
Applications includes sterile filling and dispensing systems, diagnostic and nuclear equipment, laboratory analytical instrumentation, infusion sets for parenterals and drugs, cosmetic production, food and beverage processing, cell and tissue culture transport.
Temperature resistance: –70 to +52 °C
Hardness (Shore durometer A): 72
Density: 0,90
Certifications: Meets USP class VI and FDA criteria.