Nitrile gloves, TouchNTuff® 92-500
Supplier: Ansell
Certificates
About this item
These nitrile gloves offer up to four times the puncture resistance of comparable natural latex gloves, and three times the resistance of similar neoprene gloves. They feature a proven splash resistance against hazardous chemicals.
- Non sterile, powdered, textured fingertips
- "Thin Nitrile Technology" formulation for easy donning and strong grip in wet or dry conditions
- Designed for maximum freedom of movement and minimum hand fatigue
- Contains no natural rubber proteins: Reduces the risk of Type I allergies
Suitable for chemical handling, light assembly tasks and applications in the electronics industry.
Certifications: EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5, Cat. III; AQL 1,5
Food contact approved.
Food contact approved.
Specifications
- Catalog No:
- 112-0996
- 112-0995
- 112-0993
- 112-0994
- Model:
- 92-500
- 92-500
- 92-500
- 92-500
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powdered
- Powdered
- Powdered
- Powdered
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, EN 1149-5
- Anti-static:
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +