Tubing, PTFE, Omnifit
: OMNIFIT
PTFE, white.
- Excellent chemical resistance
- Excellent thermal resistance, autoclavable
- Free of extractable substances, physiologically neutral, biocompatible
Excellent resistance to acids, bases and solvents.
PTFE, white.
Excellent resistance to acids, bases and solvents.