Order Entry
Germany
Orders LinkContactUsLinkComponent
Bed pan

Bed pan

2500 ml, 440×270×95 mm
Catalog # DELT19586
Supplier:   DELTALAB
Bed pan
Catalog # DELT19586
Supplier:  DELTALAB
Bed pan

2500 ml, 440×270×95 mm

Price
316,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

Bed pan made of white PP, designed for disabled people, for the collection of urine and faeces.

  • Features rounded edges for more comfort.
  • L-shaped handle for a better handling
  • Allows a secure one-handed grip and avoids accidental spillage
  • Can be washed in automatic machines
  • Useful for hanging on handles and beds

 : Supplied as individually wrapped and with instructions for use.

Specifications
  • Capacity:
    2500 ml
  • W×D×H:
    440×270×95 mm