Order Entry
Germany
ContactUsLinkComponent
Microtube racks, incubation
Microtube racks, incubation
Catalog # DELTM-022
Supplier:  DELTALAB
Microtube racks, incubation
Catalog # DELTM-022
Supplier:  DELTALAB

Specifications

  • For tubes:
    0,5 and 1,5 ml
  • No. of Holes:
    24
  • Description:
    Incubation rack, 119×78 mm
  • Pk:
    4

Specifications

About this item

This floating rack, made of PP, can incubate up to 24 microtubes in a water bath. Tubes are held snugly and then released when the rack is pressed
down on a benchtop.

  • Alphanumerically labelled for better sample identification
  • Autoclavable
  • Excellent chemical resistance