Orders LinkContactUsLinkComponent
Analytical HPLC columns, XTerra® MS C8
Analytical HPLC columns, XTerra® MS C8
Marketsource
Supplier:   WATERS
Analytical HPLC columns, XTerra® MS C8
Supplier:  WATERS
Marketsource
Certificates
Analytical HPLC columns, XTerra® MS C8
2 Options Available Below
Product Details & Documents
XTerra® MS C8 columns are general purpose, hybrid-based, reversed phase C8 columns that are designed to be compatible with mass spectrometry applications. These first generation columns provide superior pH stability compared to a silica based column.
Product Family Options
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    MS C8
    125 Å
    3,5 µm
    2,10 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 270,00 €
    1 270,00 €
    1 * 1 ST
       1 270,00 €
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    MS C8
    125 Å
    3,5 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 310,00 €
    1 310,00 €
    1 * 1 ST
       1 310,00 €
    Specifications:
    Marketsource
    Marketsource